Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Petroleum spirit ACS

Supplier: Ricca Chemical
Danger
Synonyms: Petroleum etherGasolinePetrol

RSOP0200-4C
RCRSOP0200-4CEA 201.65 USD InStock
RCRSOP0200-4C
Petroleum spirit ACS
Petroleum spirit


Boiling Pt: 20°C - 75°C
Density: 0.6450
Flash Pt: < -18°C
MDL Number: MFCD00081849
CAS Number: 8032-32-4
UN: 1268
ADR: 3,II
Order Now
Specification Test Results
Acidity To Pass Test
Boiling Range 35 - 60 °C
Color (APHA) max 10
Residue after evaporation (Non-Volatile Matter) max 0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty