Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene ACS

Supplier: Ricca Chemical
Danger
Synonyms: MethacideBenzene, methylMethylbenzolToluolPhenylmethaneMethylbenzeneMethane, phenyl-

RSOT0200-20E4 RSOT0200-4C RSOT0200-4X4C
RCRSOT020020E4EA 396.07 USD InStock
RCRSOT020020E4 RCRSOT0200-4C RCRSOT02004X4C
Toluene ACS
Toluene


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: ∼ 111°C
Melting Pt: -95°C
Density: 0.8660
Flash Pt: 4°C
MDL Number: MFCD00008512
CAS Number: 108-88-3
UN: 1294
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg
Order Now
Specification Test Results
Assay min 99.5 %
Color (APHA) max 10
Sulfur compounds (as S) max 0.003 %
Water (H₂O) max 0.03 %
Residue after evaporation (Non-Volatile Matter) max 0.001 %
Substances Darkened by Sulfuric Acid To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty