Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xylene (mixture of isomers) ACS

Supplier: Ricca Chemical
Danger
Synonyms: Dimethylbenzene

RSOX0200-4C RSOX0200-4X4C
RCRSOX02004X4CCS 379.93 USD InStock
RCRSOX02004X4C RCRSOX0200-4C
Xylene (mixture of isomers) ACS
Xylene (mixture of isomers)


Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 138.3 - 141.4°C
Melting Pt: -47.9 - 13.3°C
Density: 0.8700
Flash Pt: 25°C
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/44/47/704447.jpg
Order Now
Specification Test Results
Assay min 98.5 % Xylenes
Color (APHA) max 10
Sulfur compounds (as S) max 0.003 %
Water (H₂O) max 0.05 %
Residue after evaporation (Non-Volatile Matter) max 0.002 %
Substances Darkened by Sulfuric Acid To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty