Would you like to log in again?
Bismuth Standard, for AAS, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010008-500N RV010008-100N
RCRV010008100NEA 68.94 USD InStock
RCRV010008100N RCRV010008500N
Bismuth Standard, for AAS, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Bismuth Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty