Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury Standard, for AAS, VeriSpec®, Ricca Chemical Company

Supplier: RICCA Chemical Company

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VeriSpec®
RV010024-500C RV010024-100C
RCRV010024100CEA 73.32 USD InStock
RCRV010024100C RCRV010024500C
Mercury Standard, for AAS, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Mercury Standards
  • NIST traceable

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility

Caution: 1000 ppm =1 g/L= 1000 µg/ml

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty