Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VeriSpec™ Nickel (Ni) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company

Supplier: RICCA Chemical Company

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VeriSpec™
RV010037-500N RV010037-100N RV010037-1N
RCRV010037100NEA 75.79 USD InStock
RCRV010037100N RCRV010037500N RCRV010037-1N
VeriSpec™ Nickel (Ni) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Nickel Standards
Certified reference material (CRM) for AA (atomic absorption)

Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty