Would you like to log in again?
VeriSpec® Cerium (Ce) Standard for AAS 1000 ppm in 2% HNO₃
Supplier: RICCA Chemical Company
VeriSpec™
RV010070-100N RV010070-500N
RCRV010070500NEA 190.95 USD InStock
RCRV010070500N RCRV010070100N
VeriSpec® Cerium (Ce) Standard for AAS 1000 ppm in 2% HNO₃
Standards Cationic Standards Cerium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty