Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HCl

Supplier: RICCA Chemical Company

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VeriSpec®
RV010261-250N RV010261-500N RV010261-50N RV010261-100N
RCRV010261500NEA 302.18 USD InStock
RCRV010261500N RCRV010261-50N RCRV010261250N RCRV010261100N
VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HCl
Standards Cationic Standards Aluminum Standards
Certified reference material (CRM) for ICP

Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty