Would you like to log in again?
VeriSpec™ Magnesium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010292-50N RV010292-250N RV010292-100N RV010292-500N
RCRV010292100NEA 146.15 USD InStock
RCRV010292100N RCRV010292500N RCRV010292250N RCRV010292-50N
VeriSpec™ Magnesium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Magnesium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty