Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdenum Standards, for ICP, VeriSpec®, Ricca Chemical Company

Supplier: RICCA Chemical Company

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VeriSpec®
RV010294-50N RV010294-250N RV010294-100N RV010294-500N RV010396-50N RV010396-250N RV010396-500N RV010396-100N
RCRV010294250NEA 210.87 USD InStock
RCRV010294250N RCRV010294500N RCRV010396500N RCRV010294100N RCRV010396100N RCRV010396-50N RCRV010294-50N RCRV010396250N
Molybdenum Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Molybdenum Standards
  • NIST traceable

Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Caution: 1000 ppm =1 g/L= 1000 µg/ml

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty