Would you like to log in again?
VeriSpec® Tellurium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010356-250N RV010356-50N RV010356-500N RV010356-100N
RCRV010356100NEA 128.58 USD InStock
RCRV010356100N RCRV010356-50N RCRV010356500N RCRV010356250N
VeriSpec® Tellurium Standard for ICP 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Tellurium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty