Would you like to log in again?
VeriSpec™ Zirconium Standard for ICP 1000 ppm in 2% HNO₃/HF, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010362-250N RV010362-100N RV010362-50N RV010362-500N
RCRV010362100NEA 128.58 USD InStock
RCRV010362100N RCRV010362250N RCRV010362500N RCRV010362-50N
VeriSpec™ Zirconium Standard for ICP 1000 ppm in 2% HNO₃/HF, RICCA Chemical Company
Standards Cationic Standards Zirconium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty