Would you like to log in again?
VeriSpec® Cerium Standards for ICP, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec® VeriSpec™
RV010271-100N RV010373-100N RV010373-250N RV010271-50N RV010271-500N RV010373-500N RV010271-250N
RCRV010271250NEA 210.87 USD InStock
RCRV010271250N RCRV010373250N RCRV010373100N RCRV010271-50N RCRV010373500N RCRV010271100N RCRV010271500N
VeriSpec® Cerium Standards for ICP, RICCA Chemical Company
Standards Cationic Standards Cerium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty