Would you like to log in again?
VeriSpec® Rubidium Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010407-50N RV010407-500N RV010407-100N RV010407-250N
RCRV010407250NEA 579.26 USD InStock
RCRV010407250N RCRV010407-50N RCRV010407500N RCRV010407100N
VeriSpec® Rubidium Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Standards Cationic Standards Rubidium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 10 g/L= 10,000 µg/mL
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty