Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VeriSpec® Molybdenum Standard for ICP 10 g/L in 4% NH₃, Ricca Chemical Company

Supplier: RICCA Chemical Company

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VeriSpec™
RV010466-250N RV010466-50N RV010466-100N RV010466-500N
RCRV010466500NEA 449.77 USD InStock
RCRV010466500N RCRV010466100N RCRV010466250N RCRV010466-50N
VeriSpec® Molybdenum Standard for ICP 10 g/L in 4% NH₃, Ricca Chemical Company
Standards Cationic Standards Molybdenum Standards
Certified reference material (CRM) for ICP

Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty