Would you like to log in again?
Tellurium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010491-500N RV010320-250N RV010421-250N RV010320-500N RV010320-50N RV010491-100N RV010421-500N RV010320-100N RV010421-100N RV010491-50N RV010491-250N
RCRV010491500NEA 661.69 USD InStock
RCRV010491500N RCRV010421100N RCRV010320500N RCRV010320100N RCRV010491100N RCRV010421500N RCRV010491250N RCRV010320250N RCRV010421250N RCRV010320-50N RCRV010491-50N
Tellurium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Tellurium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty