Would you like to log in again?
VeriSpec® ICP-MS Tuning Solution 9 Components, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010665-100N
RCRV010665100NEA 478.3 USD InStock
RCRV010665100N
VeriSpec® ICP-MS Tuning Solution 9 Components, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
Certified reference material (CRM) for ICP-MS
10 ppm: Ba, Be, Ce, Co, In, Mg, Pb, Th, Tl
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty