Would you like to log in again?
VeriSpec® Mixed Cation Standard, 6 Components, Ricca Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010691-100N
RCRV010691100NEA 425.23 USD InStock
RCRV010691100N
VeriSpec® Mixed Cation Standard, 6 Components, Ricca Chemical Company
Standards Cationic Standards Multi-Element Standards
Certified reference material (CRM) for ion chromatography (IC)
50 ppm Li+, Na+ 200 ppm; 250 ppm NH₄⁺, Mg²⁺ ; 500 ppm K+, Ca²⁺ 500
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty