Would you like to log in again?
VeriSpec® ICP Multi-Element QC Standard 7, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010705-100N
RCRV010705100NEA 509.53 USD InStock
RCRV010705100N
VeriSpec® ICP Multi-Element QC Standard 7, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
7 component standard.
1000 ppm: K, 500 ppm Si; 100 ppm Al, B, Ba, Na
50 ppm: Ag
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty