Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VeriSpec® Strontium - Sr²⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec™
RV010941-250N RV010941-100N RV010941-500N
RCRV010941500NEA 189.94 USD InStock
RCRV010941500N RCRV010941100N RCRV010941250N
VeriSpec® Strontium - Sr²⁺ Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards Cationic Standards Strontium Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty