Would you like to log in again?
Verispec® Certified Reference Material/Standard, KCl Conductivity Standards at 25°C, RICCA Chemical Company
Supplier: RICCA Chemical Company
RV588860-500A RV588775-500A RV588684-500A RV588750-500A RV588843-500A
RCRV588860500AEA 63.3 USD InStock
RCRV588860500A RCRV588775500A RCRV588684500A RCRV588843500A RCRV588750500A
Verispec® Certified Reference Material/Standard, KCl Conductivity Standards at 25°C, RICCA Chemical Company
Standards Conductivity Standards
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: Store at room temperature. The conductivity of these certified reference materials are strongly dependent on temperature. All calibrations with these certified reference materials should be conducted at 25°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty