Would you like to log in again?
Rtx®-5MS GC Columns, Restek
Supplier: Restek
Rtx®
12621 12620-124 12620 12654 12623-124042 12626-124 12623-042 12624 12655-6850 12639 12626-105 12626 12626-127 12652-126 12623-127 12623-124 12667 12638 12620-124600 12626-6850 12635-127 12606 12624-128 12641 12638-124 12623 12642-128 12670 12605-123 12611 12605 12623-600 12608 12624-042 12608-124 12624-600 12623-112 12667-126 12627 12653 12624-125 12642 12623-305 12655 12635 12620-6850 12640 12623-124106 12637 12650 12639-125 12652 12638-127 12609 12623-6850 12653-127 12620-600
10849-688EA 536.48 USD InStock
10849-688 10849-646 75852-034 10850-132 10060-044 RK12667 RK12623 10850-328 RK12624 75851-036 10849-130 RK12620 RK12621 10850-420 10849-154 RK12623-124 10850-424 10850-346 RK12627 10803-460 10850-426 10850-140 10850-340 10852-012 RK12670 RK12638-127 10059-030 10850-418 10850-430 10849-144 10850-432 10849-468 RK12638 RK12639 10850-118 10850-428 10059-028 RK12640 10849-134 10850-364 10850-122 10849-254 RK12609 10850-124 RK12605 RK12606 10849-136 10850-326 10850-120 RK12650 10849-060 RK12611 RK12655 10849-382 75851-044 RK12653 RK12654
Rtx®-5MS GC Columns, Restek
Chromatography Columns
Low-bleed columns developed for mass spectrometers and the analysis of semi-volatile pollutants.
  • Low bleed
  • High degree of inertness
  • Internal dimensions and film thickness vary between models
The synthesis and bonding procedures for the Rtx®-5MS were modified from standard 5% phenyl polymers to address specific bleed concerns by mass spectroscopists with attention being paid to reduce bleed fragments and baseline rise
Note: The DB-5MS is a silarylene-based polymer equivalent to the Rxi-5Sil MS.
Each column is shipped with the actual test chromatograph that demonstrates the high degree of inertness for stringent environmental compounds and shows minimal baseline rise at the upper operating temperature. Also included is a test chromatogram showing the polymer bleed profile by a sensitive MS detector. Phase composition: crossbonded 5% diphenyl, 95% dimethylpolysiloxane. Temperature range: –60 to 330/350 °C (–76 to 626/662 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty