Would you like to log in again?
Micropacked GC Columns, Restek
Supplier: Restek
19002 19023 19915-6850 19042 19004 19017 19006 19014 19012 19013 19001-291 19005-900 19015 19003 19011 19001 19008 19040 19010 19020 19009 19005 19022 19021 19041 19016 19018 19007 19915-600 19000 19019
RK19011EA 223.47 USD InStock
RK19011 RK19010 RK19019 RK19018 RK19017 RK19016 RK19015 RK19014 RK19013 RK19012 10803-466 10803-204 10849-058 RK19022 RK19000 10848-096 RK19021 10847-808 RK19020 10847-806 RK19008 RK19007 RK19006 RK19005 RK19004 10848-100 RK19003 RK19002 RK19023 RK19001 RK19009
Micropacked GC Columns, Restek
Chromatography Columns
Micro-packed columns are highly efficient and provide good sample capacity, resulting in a powerful tool for solving many difficult application problems.
  • Increased efficiency over traditional packed columns
  • Higher capacity than PLOT columns
  • Made from inert, flexible SilcoSmooth® tubing
  • Wide range of packings available
  • Standard coils fit all instruments. No special instrument configurations required.
The unsurpassed inertness of SilcoSmooth® tubing is based on Siltek® deactivation, which allows the column to be flexed and coiled without any fear of chipping or cracking the inert surface.
Our micro-packed columns are designed to fit packed and capillary injection systems. Standard wall (¹/₁₆" Ext.Ø.) micro-packed columns offer improved efficiency in packed column instruments without the expense of converting to capillary injection systems. Smaller (0.95 mm Ext.Ø.) micro-packed columns install easily into a capillary injector, using slightly larger ferrules. Micro-packed columns operate at flows exceeding 10 cc/min for trouble-free operation.
Restek’s packed column experts insert braided wire into the column and secure it by making a small crimp near the column outlet. End plugs are Siltek® treated—the sample contacts only inert surfaces.
Ordering information: *Due to the small internal diameter of 0.53 mm Int.Ø. columns, braided wire end plugs cannot be used; wool is inserted into the ends instead.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty