Would you like to log in again?
Slide-Lock Tweezers and 15 cm Compact Steel Ruler, Restek
Supplier: Restek
20101
RK20101EA 39.89 USD InStock
RK20101
Slide-Lock Tweezers and 15 cm Compact Steel Ruler, Restek
Forceps
This duo provides an alternative way to install capillary columns to the correct insertion distance recommended by the instrument manufacturer.
  • Useful for many laboratory tasks
  • Durable steel construction
  • Easily install capillary columns
This duo provides an alternative way to install capillary columns to the correct insertion distance recommended by the instrument manufacturer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty