Would you like to log in again?
Pocket Magnifier, Restek
Supplier: Restek
20124
RK20124EA 49.55 USD InStock
RK20124
Pocket Magnifier, Restek
Magnifiers
10X magnification makes it easy to see the column end to verify a square cut.
  • Small and easy to handle
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty