Would you like to log in again?
Graphite Replacement Ferrules, Restek
Supplier: Restek
20210
RK20210PK 69.18 USD InStock
RK20210
Graphite Replacement Ferrules, Restek
Fittings and Connectors
Ferrules are sized to be suitable for 1/4" fittings.
  • 1/4" fitting sizing
  • Graphite fabrication
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty