Would you like to log in again?
Compact Ferrules for Agilent GCs, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Supplier: Restek
27070 20239 20238 20250 20251 27071
RK20239PK 388.3 USD InStock
RK20239 RK20238 10803-416 RK20251 RK20250 10852-544
Compact Ferrules for Agilent GCs, Fit ≤0.28 mm int.Ø Fused Silica Columns, Restek
Fittings and Connectors
Compact ferrules fit fused silica columns with internal diameters ≤0.28 mm.
  • Preconditioned to eliminate outgassing
  • High-purity, high-density graphite or Vespel®/graphite material
  • Stable to 400°C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty