Would you like to log in again?
Two-Hole Ferrules for ¹/₈ and ¹/₁₆" Compression-Type Fittings, Restek
Supplier: Restek
24849 24848 20246
10059-312PK 81.77 USD InStock
10059-312 10059-310 RK20246
Two-Hole Ferrules for ¹/₈ and ¹/₁₆" Compression-Type Fittings, Restek
Fittings and Connectors
Two-hole ferrules seal with minimal torque, are reusable, and are preferred for vacuum and high-pressure uses.
  • Use 1/16-inch, two-hole ferrules with the 1/16-inch capillary inlet adaptor fitting kit
  • Use 1/8-inch, two-hole ferrules with the 1/8-inch capillary inlet adaptor fitting kit
  • Stable to 400°C
For use with 1/8" and 1/16" compression-type fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty