Would you like to log in again?
MXT® Low Dead Volume Connector Kits for Metal Columns, Restek
Supplier: Restek
MXT®
20394 20536 20397
10852-986EA 98.93 USD InStock
10852-986 10852-988 RK20397
MXT® Low Dead Volume Connector Kits for Metal Columns, Restek
Fittings and Connectors
MXT® low dead volume connector kits for metal columns can be used to connect a guard column or transfer line to an MXT® stainless steel column.
  • Connect a guard column/transfer line to an MXT® stainless steel column
  • Low thermal mass tracks rapid oven temperature programming
  • Stainless steel ferrules and nuts
  • Available in “Y” and union configurations
  • Siltek® treatment ensures ultimate inertness
Each kit contains the MXT® union, two 1/32-inch ferrules and nuts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty