Would you like to log in again?
¹/₈" Capillary Inlet Adapter Fitting Kit, Split/Splitless Fitting for 0.53 mm Int.Ø Capillary Columns, Restek
Supplier: Restek
20645
RK20645EA 122.78 USD InStock
RK20645
¹/₈" Capillary Inlet Adapter Fitting Kit, Split/Splitless Fitting for 0.53 mm Int.Ø Capillary Columns, Restek
Fittings and Connectors
1/8-inch split/splitless fitting accepts standard two-hole capillary ferrules and a standard 1/8-inch nut.
  • Makes column installation easy due to the nut protruding farther from the insulated injection port chamber
  • The column insertion depth is the same as the original equipment
  • Kit includes adapter fitting, capillary nut, stainless steel inlet seal, washer, and one 0.8 mm ID two-hole ferrule
  • Use recessed taper liners with this adapter
These inlet adapter fitting kits make column installation easy with far-protruding nuts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty