Would you like to log in again?
Double Taper Inlet Liner for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets, Restek
Supplier: Restek
20953 20953-214.5
RK20953PK 251.72 USD InStock
RK20953 75779-034
Double Taper Inlet Liner for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty