Would you like to log in again?
Finger-Tight Capillary Column Nuts, Restek
Supplier: Restek
21311 21312
RK21311EA 51.69 USD InStock
RK21311 RK21312
Finger-Tight Capillary Column Nuts, Restek
Fittings and Connectors
Rapidly tighten without wrenches; avoid stripped threads.
  • Can be used with 0.25, 0.32, or 0.53 mm ID columns
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty