Would you like to log in again?
Dual-Stage Ultra-High Purity Chrome-Plated Brass Gas Regulators with CGA Fittings, Restek
Supplier: Restek
21667 21668 21669
RK21668EA 1182.34 USD InStock
RK21668 RK21669 RK21667
Dual-Stage Ultra-High Purity Chrome-Plated Brass Gas Regulators with CGA Fittings, Restek
Gas Regulators
Oxidation-resistant, chrome-plated dual-stage brass gas regulators are ideal for sensitive GC applications using MS, PID, or ECD detection methods.
  • Most stable outlet pressure control
  • Secondary pressure regulation not needed
  • Most widely used regulator
  • Less internal volume than stainless steel gas regulators
Regulators feature CGA fittings, and are also available with international fittings. Custom gas regulators are also available.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty