Would you like to log in again?
Injector/Detector Plug Nuts, Restek
Supplier: Restek
21883
RK21883PK 58.92 USD InStock
RK21883
Injector/Detector Plug Nuts, Restek
Fittings and Connectors
Use injector/detector plug nuts for a variety of purposes, including:
  • Use to cap off an injector to isolate leaks.
  • Use to cap off a detector for thermal cleaning.
  • Use to check a detector or make-up gas flow rate.
  • Use to cap off a detector and prevent hydrogen from accidentally diffusing into the oven from an unused detector base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty