Would you like to log in again?
Wall Mounting Bracket, Restek
Supplier: Restek
21984
RK21984EA 154.44 USD InStock
RK21984
Wall Mounting Bracket, Restek
Wall Hooks
Optional wall mounting bracket allows for the mounting of Super Clean® base plates with the use of screws.
  • Additional support for mounting base plates
Base plates can be mounted by using screws and the mounting holes on the base plate, or by using this optional wall mounting bracket.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty