Would you like to log in again?
Capillary-Grade Hydrocarbon Trap, Restek
Supplier: Restek
21992 21991
RK21992EA 254.25 USD InStock
RK21992 RK21991
Capillary-Grade Hydrocarbon Trap, Restek
Filters Gas Filters
Packed with an extremely high surface area, baked coconut shell-based activated carbon.
  • Purged with ultra-high-purity helium; ready to use with any carrier gas including hydrogen
  • Reduces organics to 0.1 ppm (assuming 100 ppm input)
  • Maximum operating pressure: 250 psi (1724 kPa)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty