Would you like to log in again?
9 mm Short-Cap, Screw-Vial Closures, Polypropylene, Preassembled, Restek
Supplier: Restek
24668 26599 24495 24496 26572 24503 24493 24488 24669 24498 26581 26580 24504 24485 26573 24487 24501 26577 26576 24497 24494 24502 26574 24500 24506 26578 26575 24505 24474 24670 24473 24486 26598
10853-966PK 51.7 USD InStock
10853-966 10853-964 RK24486 RK24485 10853-968 10854-310 10854-312 10059-164 10854-306 10854-308 10853-892 10059-168 10059-166 10853-970 10853-958 RK24474 10853-956 10854-262 10854-264 10854-384 RK24498 10854-266 10854-320 10854-322 10854-314 10854-316 10059-172 10059-170 10854-318 10803-356 10853-962 10853-960 10803-358
9 mm Short-Cap, Screw-Vial Closures, Polypropylene, Preassembled, Restek
Closures Chromatography Vial Closures
9 mm short-cap closures are compatible with screw-thread or ribbed designs.
  • Economical
  • Sturdy polypropylene construction
These screw caps are for use with vials featuring short external threads where standard-length caps would be too long.
Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty