Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High-Pressure Cup-Type In-Line Filter, Restek

Supplier: Restek

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

25000 25001
RK25001EA 195.09 USD InStock
RK25001 RK25000
High-Pressure Cup-Type In-Line Filter, Restek
Filters In-Line Filters
This high-pressure cup-type filter can be used in fluid streams operating to 15,000 psi (103,421 kPa).

The cup-shaped filter elements have a large (2.5 cm²) surface area to give long operating lifetime. Mounted in screw-type adapters, they are easily removed for cleaning. Normally, backflushing and cleaning in an ultrasonic bath with an appropriate solvent will restore them. If they become permanently clogged, replacement elements are available. Housings and all wetted parts are 316 stainless steel. Filters are packaged with gland nuts and ferrules. A bulkhead type fitting is available for thru-panel mounting. Tubing ¹/₁₆" OD; connectors–CPI.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty