Would you like to log in again?
LO-Pulse Pulse Dampers, Restek
Supplier: Restek
25012 25013
RK25013EA 508 USD InStock
RK25013 RK25012
LO-Pulse Pulse Dampers, Restek
Flow Meters
The LO-Pulse Pulse Dampers are patented, wide-dynamic-range 316 stainless steel device that smooths pulsations and maintains constant flow at system pressures from 500 to 6000 psi (3447 to 41369 kPa).
The flow path volume is only 0.9 ml and the path are efficiently swept, eliminating solvent memory effects when changing mobile phases.
Delivery information: The LO-Pulse pulse damper kit version is a space-saving, economical kit that includes hardware for mounting the pulse damper on a bracket, or for installing feet on it for bench-top use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty