Would you like to log in again?
Resprep® SPE Cartridges, Restek
Supplier: Restek
Resprep®
26063
RK26063PK 1119.49 USD InStock
RK26063
Resprep® SPE Cartridges, Restek
Chromatography Columns
Resprep® SPE cartridges contain hydrophilic adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents.
  • Constructed of PTFE frits or glass tubes with PTFE frits
EPA method 548.1 is specifically designed to provide consistent and reproducible results for the extraction of endothal from aqueous samples. Weak anion exchange resin packing (BioRex 5).
Packaging: Packed with weak anion exchange resin (BioRex 5).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty