Would you like to log in again?
Fluorobenzene, Standard, 2000 μg/ml, Restek
Supplier: Restek
30030
RK30030EA 45.59 USD InStock
RK30030
Fluorobenzene, Standard, 2000 μg/ml, Restek
Standards Organic Standards
EPA method 624 surrogate standard fluorobenzene.
  • 2000 μg/mL in P&T methanol, 1 mL/ampoule
Packaged in purge-and-trap-grade methanol at 2000 μg/mL in P&T methanol. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty