Would you like to log in again?
EPA method 624 Standards Kit, Restek
Supplier: Restek
30055
RK30055EA 228.92 USD InStock
RK30055
EPA method 624 Standards Kit, Restek
Standards Environmental Analysis Standards
Calibration mixes are packaged in purge and trap methanol at a concentration of 2000 µg/mL in a 1 mL ampoule.
  • Contents of 624 Calibration Mixture No. 2: Benzene, Carbon Tetrachloride, Chlorobenzene, 2-Chloroethyl Vinyl Ether, Dibromochloromethane, 1,1-Dichloroethane, 1,1-Dichloroethene, 1,2-Dichloropropane, Methylene Chloride, Tetrachloroethylene, 1,1,2-Trichloroethane, and Trichloroethene
Internal standard mix is packaged in purge and trap methanol at a concentration of 1500 µg/mL in a 1 mL ampoule. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty