Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
8011 Calibration Mix—EDB/DBCP (2 components), Restek
Supplier: Restek
30062
RK30062EA 36.86 USD InStock
RK30062
8011 Calibration Mix—EDB/DBCP (2 components), Restek
Standards Environmental Analysis Standards
Mix contains: 1,2-Dibromo-3-chloropropane (DBCP) (96-12-8), 1,2-Dibromoethane (EDB) (106-93-4).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty