Would you like to log in again?
UST Gasoline Range Organics (GRO) Standard, Restek
Supplier: Restek
30065
RK30065EA 43.65 USD InStock
RK30065
UST Gasoline Range Organics (GRO) Standard, Restek
Standards Petroleum Industry Standards
Standards are packaged in purge-and-trap-grade methanol at the stated concentrations in a 1 mL ampoule.
  • Mixture contains 500 µg/mL Benzene, 500 µg/mL Ethylbenzene, 500 µg/mL Heptane, 1500 µg/mL 2-Methylpentane, 1500 µg/mL Toluene, 1000 µg/mL 1,2,4-Trimethylpentane, 1500 µg/mL 2,2,4-Trimethylpentane, 1000 µg/mL m-Xylene, and 1000 µg/mL o-Xylene
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty