Would you like to log in again?
EPA method 8260 Volatile Organics Standards Kit, Restek
Supplier: Restek
30076
RK30076EA 408.37 USD InStock
RK30076
EPA method 8260 Volatile Organics Standards Kit, Restek
Standards Environmental Analysis Standards
Standards are packaged at a concentration of 2500 µg/mL (except 2000 µg/mL for 502.2 calibration mixes and system performance check mix) in purge-and-trap-grade methanol at a concentration in 1 mL ampoules.
  • Contents of 502.2 Calibration Mixture No. 1: Bromomethane, Chloromethane, Chloroethane, Dichlorodifluoromethane, Trichlorofluoromethane, Vinyl Chloride.
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty