Would you like to log in again?
L/C VOA Internal Standard Mix (3 Components), Restek
Supplier: Restek
30091
RK30091EA 45.59 USD InStock
RK30091
L/C VOA Internal Standard Mix (3 Components), Restek
Standards Organic Standards
Mix contains: Chlorobenzene-d5 (3114-55-4), 1,4-Dichlorobenzene-d4 (3855-82-1), 1,4-Difluorobenzene (540-36-3).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty