Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
524 Rev. 4.0 Volatile Organics Kit (2,000 µg/mL), Restek
Supplier: Restek
30204
RK30204EA 387.03 USD InStock
RK30204
524 Rev. 4.0 Volatile Organics Kit (2,000 µg/mL), Restek
Standards Environmental Analysis Standards
Contains 1 ml each of these mixtures: 524 internal standard/surrogate mix, 502.2 calibration mix #1, 502.2 calibration mix #2, 502.2 calibration mix #3, 502.2 calibration mix #4, 502.2 calibration mix #5, 502.2 calibration mix #6, 524 calibration mix #7a, 524 calibration mix #7b, 524 calibration mix #8.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty