Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
8240 Alcohols Mix (5 components), Restek
Supplier: Restek
30214
RK30214EA 34.92 USD InStock
RK30214
8240 Alcohols Mix (5 components), Restek
Standards Environmental Analysis Standards
Mix contains: Allyl alcohol (2-propen-1-ol) (107-18-6); 2-Chloroethanol (107-07-3); Ethanol (64-17-5); Isobutyl alcohol (78-83-1); Propargyl alcohol (107-19-7).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty