Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
DW-VOC Kit #2 (2,000 µg/ml), Restek
Supplier: Restek
30221
RK30221EA 153.26 USD InStock
RK30221
DW-VOC Kit #2 (2,000 µg/ml), Restek
Standards Environmental Analysis Standards
Contains: 30211: 501 Trihalomethane Mix; Bromodichloromethane (75-27-4); Bromoform (75-25-2); Chloroform (67-66-3); Dibromochloromethane (124-48-1);30219: DW-VOC Mix #1; Benzene (71-43-2); Carbon tetrachloride (56-23-5); 1,4-Dichlorobenzene (106-46-7); 1,2-Dichloroethane (107-06-2); 1,1-Dichloroethene (75-35-4); 1,1,1-Trichloroethane (71-55-6); Trichloroethene (79-01-6); Vinyl chloride (75-01-4);30220: DW-VOC Mix #2; Chlorobenzene (108-90-7); 1,2-Dichlorobenzene (95-50-1); cis-1,2-Dichloroethene (156-59-2); trans-1,2-Dichloroethene (156-60-5); 1,2-Dichloropropane (78-87-5); Ethylbenzene (100-41-4); Styrene (100-42-5); Tetrachloroethene (127-18-4); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3);30235: DW-VOC Mix #3; Methylene chloride (dichloromethane) (75-09-2); 1,2,4-Trichlorobenzene (120-82-1); 1,1,2-Trichloroethane (79-00-5)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty