Would you like to log in again?
PVOC Mix (California) (7 Components), Restek
Supplier: Restek
30231
RK30231EA 39.77 USD InStock
RK30231
PVOC Mix (California) (7 Components), Restek
Standards Environmental Analysis Standards
Mix contains: Benzene (71-43-2); Ethylbenzene (100-41-4); Methyl tert-butyl ether (MTBE) (1634-04-4); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty